Product Details

SNP ID
rs1370276
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.15:80157986 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGGGACCTGTGGACTCTTCAATAGA[C/T]AGGCTTTCTGAGTAAATGAGCCAAG
Phenotype
MIM: 613871
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAH PubMed Links

Gene Details

Gene
FAH
Gene Name
fumarylacetoacetate hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000137.2 Intron NP_000128.1

View Full Product Details