Product Details

SNP ID
rs1972435
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.15:89686090 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATAACTATTCCACTCTTGTTTAGAA[C/T]AAAGGGGCTCACAGGAGCAGAAGCT
Phenotype
MIM: 603866 MIM: 170290
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PEX11A PubMed Links

Gene Details

Gene
PEX11A
Gene Name
peroxisomal biogenesis factor 11 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271572.1 Intron NP_001258501.1
NM_001271573.1 Intron NP_001258502.1
NM_003847.2 Intron NP_003838.1
Gene
PLIN1
Gene Name
perilipin 1
There are no transcripts associated with this gene.

Gene
WDR93
Gene Name
WD repeat domain 93
There are no transcripts associated with this gene.

View Full Product Details