Product Details

SNP ID
rs1862896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:95518675 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATTGTTTTTGACATGTAGGTAAC[C/A]CATACTTTTCCAAAATATGTGCATC
Phenotype
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
LOC100419920 PubMed Links

Gene Details

Gene
LOC100419920
Gene Name
ankyrin repeat domain 33B pseudogene
There are no transcripts associated with this gene.

Gene
TRIM64FP
Gene Name
tripartite motif containing 64F, pseudogene
There are no transcripts associated with this gene.

View Full Product Details