Product Details

SNP ID
rs1878145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:183120573 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTGAAGAGGCAGAAAGAATATCT[C/G]AATGTTTGATGGTAGAGAAGGTGAA
Phenotype
MIM: 608140
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NUP35 PubMed Links

Gene Details

Gene
NUP35
Gene Name
nucleoporin 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287584.1 Intron NP_001274513.1
NM_001287585.1 Intron NP_001274514.1
NM_138285.4 Intron NP_612142.2
XM_006712254.2 Intron XP_006712317.1
XM_011510576.2 Intron XP_011508878.1
XM_011510577.2 Intron XP_011508879.1
XM_017003308.1 Intron XP_016858797.1

View Full Product Details