Product Details

SNP ID
rs2003752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23399918 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCTGTAGCACAGCGTAGGACCTC[A/G]GAAGCCCTCAGACAGCACACTGGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC107985555 PubMed Links

Gene Details

Gene
LOC107985555
Gene Name
coiled-coil domain-containing protein 188-like
There are no transcripts associated with this gene.

Gene
ZDHHC8P1
Gene Name
zinc finger DHHC-type containing 8 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details