Product Details

SNP ID
rs14075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7235285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAAAAGATGAGGGAGGGGAGTG[C/T]TAATATTTACACTAGAGTTTTATAG
Phenotype
MIM: 610684 MIM: 602151 MIM: 605125 MIM: 612910
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTDNEP1 PubMed Links

Gene Details

Gene
CTDNEP1
Gene Name
CTD nuclear envelope phosphatase 1
There are no transcripts associated with this gene.

Gene
DVL2
Gene Name
dishevelled segment polarity protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004422.2 1579 Intron NP_004413.1
XM_005256502.1 1579 Intron XP_005256559.1
XM_017024298.1 1579 Intron XP_016879787.1
Gene
GABARAP
Gene Name
GABA type A receptor-associated protein
There are no transcripts associated with this gene.

Gene
PHF23
Gene Name
PHD finger protein 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284517.1 1579 UTR 3 NP_001271446.1
NM_001284518.1 1579 UTR 3 NP_001271447.1
NM_024297.2 1579 UTR 3 NP_077273.2

View Full Product Details