Product Details

SNP ID
rs1883659
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:4610675 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGTTACCACAAAACCAAACCCATCA[A/G]AAGTTTGCAGGACGTCCTTGTTTAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KU-MEL-3 PubMed Links

Gene Details

Gene
KU-MEL-3
Gene Name
uncharacterized LOC497048
There are no transcripts associated with this gene.

Gene
LOC105374896
Gene Name
uncharacterized LOC105374896
There are no transcripts associated with this gene.

View Full Product Details