Product Details

SNP ID
rs15505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:24119175 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATAGTCTATCACTATCAAAAGAA[A/G]CACTATGCTAATATTTCCATATTAT
Phenotype
MIM: 190160
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
THRB PubMed Links

Gene Details

Gene
THRB
Gene Name
thyroid hormone receptor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000461.4 5517 UTR 3 NP_000452.2
NM_001128176.2 5517 UTR 3 NP_001121648.1
NM_001128177.1 5517 UTR 3 NP_001121649.1
NM_001252634.1 5517 UTR 3 NP_001239563.1
XM_005265421.4 5517 Intron XP_005265478.1
XM_005265423.4 5517 Intron XP_005265480.1
XM_005265424.3 5517 Intron XP_005265481.1
XM_006713317.3 5517 UTR 3 XP_006713380.1
XM_006713318.3 5517 Intron XP_006713381.1
XM_011534046.2 5517 Intron XP_011532348.1
XM_011534047.2 5517 Intron XP_011532349.1
XM_011534048.2 5517 Intron XP_011532350.1
XM_011534049.2 5517 Intron XP_011532351.1
XM_011534050.2 5517 Intron XP_011532352.1
XM_011534051.2 5517 Intron XP_011532353.1
XM_011534052.2 5517 Intron XP_011532354.1
XM_011534053.2 5517 Intron XP_011532355.1
XM_017007108.1 5517 Intron XP_016862597.1
XM_017007109.1 5517 Intron XP_016862598.1
XM_017007110.1 5517 Intron XP_016862599.1
XM_017007111.1 5517 Intron XP_016862600.1
XM_017007112.1 5517 Intron XP_016862601.1
XM_017007113.1 5517 Intron XP_016862602.1
XM_017007114.1 5517 Intron XP_016862603.1
XM_017007115.1 5517 Intron XP_016862604.1

View Full Product Details