Product Details

SNP ID
rs14644
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49108181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGGTCAGAAGAATTGAAGGTACA[A/T]CAGAATCAAATCACGCAGCACTGGA
Phenotype
MIM: 603727 MIM: 614471
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR6890 PubMed Links

Gene Details

Gene
MIR6890
Gene Name
microRNA 6890
There are no transcripts associated with this gene.

Gene
QARS
Gene Name
glutaminyl-tRNA synthetase
There are no transcripts associated with this gene.

Gene
USP19
Gene Name
ubiquitin specific peptidase 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199160.1 4649 Intron NP_001186089.1
NM_001199161.1 4649 UTR 3 NP_001186090.1
NM_001199162.1 4649 UTR 3 NP_001186091.1
NM_006677.2 4649 Intron NP_006668.1
XM_005264823.2 4649 Intron XP_005264880.1
XM_005264824.2 4649 Intron XP_005264881.1
XM_005264825.2 4649 Intron XP_005264882.1
XM_005264826.2 4649 Intron XP_005264883.1
XM_005264827.2 4649 Intron XP_005264884.1
XM_005264829.2 4649 UTR 3 XP_005264886.1
XM_005264830.2 4649 Intron XP_005264887.1
XM_005264831.2 4649 UTR 3 XP_005264888.1
XM_006712946.2 4649 Intron XP_006713009.1
XM_006712947.2 4649 Intron XP_006713010.1
XM_006712948.2 4649 UTR 3 XP_006713011.1
XM_006712949.2 4649 Intron XP_006713012.1
XM_006712950.2 4649 UTR 3 XP_006713013.1
XM_006712951.2 4649 UTR 3 XP_006713014.1
XM_006712952.2 4649 UTR 3 XP_006713015.1
XM_006712953.1 4649 Intron XP_006713016.1
XM_017005615.1 4649 Intron XP_016861104.1
XM_017005616.1 4649 Intron XP_016861105.1
XM_017005617.1 4649 Intron XP_016861106.1
XM_017005618.1 4649 UTR 3 XP_016861107.1
XM_017005619.1 4649 UTR 3 XP_016861108.1
XM_017005620.1 4649 UTR 3 XP_016861109.1
XM_017005621.1 4649 Intron XP_016861110.1
XM_017005622.1 4649 UTR 3 XP_016861111.1
XM_017005623.1 4649 Intron XP_016861112.1
XM_017005624.1 4649 UTR 3 XP_016861113.1
XM_017005625.1 4649 Intron XP_016861114.1
XM_017005626.1 4649 Intron XP_016861115.1
XM_017005627.1 4649 Intron XP_016861116.1
XM_017005628.1 4649 Intron XP_016861117.1
XM_017005629.1 4649 Intron XP_016861118.1
XM_017005630.1 4649 UTR 3 XP_016861119.1
XM_017005631.1 4649 UTR 3 XP_016861120.1
XM_017005632.1 4649 UTR 3 XP_016861121.1
XM_017005633.1 4649 UTR 3 XP_016861122.1
XM_017005634.1 4649 UTR 3 XP_016861123.1
XM_017005635.1 4649 Intron XP_016861124.1

View Full Product Details