Product Details

SNP ID
rs2055886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:25317160 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACAAATAGGTTTTTACTTTTATGG[A/C]AGCTCCTTCATTTTACCTGGGCTGC
Phenotype
MIM: 611792
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZCCHC4 PubMed Links

Gene Details

Gene
ZCCHC4
Gene Name
zinc finger CCHC-type containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318148.1 Intron NP_001305077.1
NM_024936.2 Intron NP_079212.2
XM_011513835.1 Intron XP_011512137.1
XM_017008128.1 Intron XP_016863617.1
XM_017008129.1 Intron XP_016863618.1

View Full Product Details