Product Details

SNP ID
rs2053835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6932225 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAGCTCAAGCCTGAGAATTACTGC[A/G]ATCTAGTAAGGCGGATTTGAACTAC
Phenotype
MIM: 605016
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF215 PubMed Links

Gene Details

Gene
ZNF215
Gene Name
zinc finger protein 215
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013250.2 581 UTR 5 NP_037382.2
XM_006718311.2 581 UTR 5 XP_006718374.1

View Full Product Details