Product Details

SNP ID
rs8677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:60028485 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATGGTTTTTCATACATATGTCAT[A/T]AGAGCTATGTGTCAATGAATGCTGA
Phenotype
MIM: 600465
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ANK3 PubMed Links

Gene Details

Gene
ANK3
Gene Name
ankyrin 3, node of Ranvier (ankyrin G)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001149.3 9452 UTR 3 NP_001140.2
NM_001204403.1 9452 UTR 3 NP_001191332.1
NM_001204404.1 9452 UTR 3 NP_001191333.1
NM_001320874.1 9452 UTR 3 NP_001307803.1
NM_020987.4 9452 UTR 3 NP_066267.2
XM_005269715.3 9452 UTR 3 XP_005269772.1
XM_006717796.3 9452 UTR 3 XP_006717859.1
XM_006717802.3 9452 UTR 3 XP_006717865.1
XM_011539708.2 9452 UTR 3 XP_011538010.1
XM_011539709.2 9452 UTR 3 XP_011538011.1
XM_017016110.1 9452 UTR 3 XP_016871599.1
XM_017016111.1 9452 UTR 3 XP_016871600.1
XM_017016112.1 9452 UTR 3 XP_016871601.1
XM_017016113.1 9452 UTR 3 XP_016871602.1
XM_017016114.1 9452 UTR 3 XP_016871603.1
XM_017016115.1 9452 UTR 3 XP_016871604.1
XM_017016116.1 9452 UTR 3 XP_016871605.1
XM_017016117.1 9452 UTR 3 XP_016871606.1
XM_017016118.1 9452 UTR 3 XP_016871607.1
XM_017016119.1 9452 UTR 3 XP_016871608.1
XM_017016120.1 9452 UTR 3 XP_016871609.1
XM_017016121.1 9452 UTR 3 XP_016871610.1
XM_017016122.1 9452 UTR 3 XP_016871611.1
XM_017016123.1 9452 UTR 3 XP_016871612.1
XM_017016124.1 9452 UTR 3 XP_016871613.1
XM_017016125.1 9452 UTR 3 XP_016871614.1
XM_017016126.1 9452 UTR 3 XP_016871615.1
XM_017016127.1 9452 UTR 3 XP_016871616.1
XM_017016128.1 9452 UTR 3 XP_016871617.1
XM_017016129.1 9452 UTR 3 XP_016871618.1
XM_017016130.1 9452 UTR 3 XP_016871619.1
XM_017016131.1 9452 UTR 3 XP_016871620.1
XM_017016132.1 9452 UTR 3 XP_016871621.1
XM_017016133.1 9452 UTR 3 XP_016871622.1
XM_017016134.1 9452 UTR 3 XP_016871623.1
XM_017016135.1 9452 UTR 3 XP_016871624.1
XM_017016136.1 9452 UTR 3 XP_016871625.1
XM_017016137.1 9452 UTR 3 XP_016871626.1
XM_017016138.1 9452 UTR 3 XP_016871627.1
XM_017016139.1 9452 UTR 3 XP_016871628.1
XM_017016140.1 9452 UTR 3 XP_016871629.1
XM_017016141.1 9452 UTR 3 XP_016871630.1
XM_017016142.1 9452 UTR 3 XP_016871631.1
XM_017016143.1 9452 UTR 3 XP_016871632.1
XM_017016144.1 9452 UTR 3 XP_016871633.1
XM_017016145.1 9452 UTR 3 XP_016871634.1
XM_017016146.1 9452 UTR 3 XP_016871635.1
XM_017016147.1 9452 UTR 3 XP_016871636.1
XM_017016148.1 9452 UTR 3 XP_016871637.1
XM_017016149.1 9452 UTR 3 XP_016871638.1
XM_017016150.1 9452 UTR 3 XP_016871639.1

View Full Product Details