Product Details
- SNP ID
-
rs1801833
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:81117553 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCCCACCATCCCTCTCCCTGTAGG[C/T]AGGAGCCCCCCAACACTGACTGCGG
- Phenotype
-
MIM: 605276
MIM: 605475
MIM: 614059
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AATK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs200561995] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AATK
- Gene Name
- apoptosis associated tyrosine kinase
- Gene
- BAIAP2
- Gene Name
- BAI1 associated protein 2
- Gene
- MIR3065
- Gene Name
- microRNA 3065
There are no transcripts associated with this gene.
- Gene
- MIR338
- Gene Name
- microRNA 338
There are no transcripts associated with this gene.
- Gene
- MIR657
- Gene Name
- microRNA 657
There are no transcripts associated with this gene.
View Full Product Details