Product Details

SNP ID
rs2063033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81167323 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTCCTGGAGACTTGCAGTTGAGT[C/T]TGATGCACCCTCATCCACTCATTCA
Phenotype
MIM: 605276
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AATK PubMed Links

Gene Details

Gene
AATK
Gene Name
apoptosis associated tyrosine kinase
There are no transcripts associated with this gene.

Gene
AATK-AS1
Gene Name
AATK antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details