Product Details

SNP ID
rs1871234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81397660 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCGGGCAGCCCCTCCCCTGACCC[C/G]TGCGCTCGCAGCCCCCTCGCCAGAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BAHCC1 PubMed Links

Gene Details

Gene
BAHCC1
Gene Name
BAH domain and coiled-coil containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291324.1 Intron NP_001278253.1
XM_011525063.2 Intron XP_011523365.1
XM_017024898.1 Intron XP_016880387.1
XM_017024899.1 Intron XP_016880388.1
Gene
LOC100130370
Gene Name
uncharacterized LOC100130370
There are no transcripts associated with this gene.

Gene
MIR4740
Gene Name
microRNA 4740
There are no transcripts associated with this gene.

View Full Product Details