Product Details
- SNP ID
-
rs1984559
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:134921318 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATACTTTTGTACTTCCAATAAATG[A/G]TCTAGCCACAAGTCTTCACTTCTTT
- Phenotype
-
MIM: 603862
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCNT2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs556347492] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCNT2
- Gene Name
- cyclin T2
- Gene
- CCNT2-AS1
- Gene Name
- CCNT2 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details