Product Details

SNP ID
rs1937301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:106820219 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGAAATATTCTTCAAGTCAAAGC[A/G]ACTAGGGCTAATAATGTTTAATCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TBC1D8B PubMed Links

Gene Details

Gene
TBC1D8B
Gene Name
TBC1 domain family member 8B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017752.2 Intron NP_060222.2
NM_198881.1 Intron NP_942582.1
XM_006724669.1 Intron XP_006724732.1
XM_006724670.3 Intron XP_006724733.1

View Full Product Details