Product Details

SNP ID
rs1806482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:67547759 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTGTGAGCCACTGCGCCCATCCCG[A/G]TTTTCTTTTGAAACCTTCTTTAGGT
Phenotype
MIM: 602520
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAP2K5 PubMed Links

Gene Details

Gene
MAP2K5
Gene Name
mitogen-activated protein kinase kinase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206804.1 Intron NP_001193733.1
NM_002757.3 Intron NP_002748.1
NM_145160.2 Intron NP_660143.1
XM_011521787.2 Intron XP_011520089.1
XM_011521788.2 Intron XP_011520090.1
XM_017022414.1 Intron XP_016877903.1

View Full Product Details