Product Details

SNP ID
rs9430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:20201851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGTCCCAGCATTCACTTCTCACA[C/T]GGGCTTCTCCAACGTGGCAGCGGCC
Phenotype
MIM: 186355
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SDC1 PubMed Links

Gene Details

Gene
SDC1
Gene Name
syndecan 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006946.1 2239 UTR 3 NP_001006947.1
NM_002997.4 2239 UTR 3 NP_002988.3
XM_005262620.4 2239 UTR 3 XP_005262677.1
XM_005262621.3 2239 UTR 3 XP_005262678.3
XM_005262622.2 2239 UTR 3 XP_005262679.1

View Full Product Details