Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
XM_011516745.2 | 1194 | Missense Mutation | ACC,TCC | T83S | XP_011515047.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_021148.2 | 1194 | Intron | NP_066971.2 | ||
XM_011515746.2 | 1194 | Intron | XP_011514048.1 | ||
XM_017011688.1 | 1194 | Intron | XP_016867177.1 | ||
XM_017011689.1 | 1194 | UTR 5 | XP_016867178.1 | ||
XM_017011690.1 | 1194 | UTR 5 | XP_016867179.1 | ||
XM_017011691.1 | 1194 | UTR 5 | XP_016867180.1 | ||
XM_017011692.1 | 1194 | UTR 5 | XP_016867181.1 | ||
XM_017011693.1 | 1194 | Intron | XP_016867182.1 | ||
XM_017011694.1 | 1194 | Intron | XP_016867183.1 | ||
XM_017011695.1 | 1194 | Intron | XP_016867184.1 | ||
XM_017011696.1 | 1194 | Intron | XP_016867185.1 | ||
XM_017011697.1 | 1194 | UTR 5 | XP_016867186.1 | ||
XM_017011698.1 | 1194 | Intron | XP_016867187.1 |