Product Details

SNP ID
rs2005080
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:22552644 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CATCCTCCAAGAGAGCAGTTCTCAC[C/T]GAGCACGAGGTGGGCGGGGAGGTCA
Phenotype
MIM: 610795
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SORBS3 PubMed Links

Gene Details

Gene
SORBS3
Gene Name
sorbin and SH3 domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018003.2 Intron NP_001018003.1
NM_005775.4 Intron NP_005766.3
XM_005273371.1 Intron XP_005273428.1
XM_006716266.1 Intron XP_006716329.1
XM_006716268.2 Intron XP_006716331.1
XM_017012944.1 Intron XP_016868433.1
XM_017012945.1 Intron XP_016868434.1
XM_017012946.1 Intron XP_016868435.1

View Full Product Details