Product Details

SNP ID
rs2247863
Assay Type
Functionally tested
NCBI dbSNP Submissions
72
Location
Chr.1:17526611 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCCAACTCCCTGTTTCTGACAAA[G/T]AGACAAGGCTGGCCGGGCACAGTCA
Phenotype
MIM: 612494
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARHGEF10L PubMed Links

Gene Details

Gene
ARHGEF10L
Gene Name
Rho guanine nucleotide exchange factor 10 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011722.2 Intron NP_001011722.2
NM_001319837.1 Intron NP_001306766.1
NM_001319838.1 Intron NP_001306767.1
NM_001328124.1 Intron NP_001315053.1
NM_018125.3 Intron NP_060595.3
XM_005245923.2 Intron XP_005245980.2
XM_005245925.2 Intron XP_005245982.2
XM_005245927.2 Intron XP_005245984.2
XM_005245929.2 Intron XP_005245986.2
XM_006710728.1 Intron XP_006710791.1
XM_006710729.1 Intron XP_006710792.1
XM_006710731.1 Intron XP_006710794.1
XM_011541691.2 Intron XP_011539993.1
XM_011541692.2 Intron XP_011539994.1
XM_011541693.2 Intron XP_011539995.1
XM_011541695.2 Intron XP_011539997.1
XM_011541696.2 Intron XP_011539998.1
XM_017001617.1 Intron XP_016857106.1
XM_017001618.1 Intron XP_016857107.1
XM_017001619.1 Intron XP_016857108.1
XM_017001620.1 Intron XP_016857109.1
XM_017001621.1 Intron XP_016857110.1
XM_017001622.1 Intron XP_016857111.1

View Full Product Details