Product Details
- SNP ID
-
hCV15754819
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:73716876 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTAATTTTTTGAAATACTGAGAAT[A/C]CAACATTTGTATGCCACACTCTACT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
BMS1P4
PubMed Links
Gene Details
- Gene
- BMS1P4
- Gene Name
- BMS1, ribosome biogenesis factor pseudogene 4
There are no transcripts associated with this gene.
View Full Product Details