Product Details
- SNP ID
-
rs2302452
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:209773024 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCATCCTGTCTTATTCATTGAAAAA[C/T]ACGTCACAAGGATGCTTTAATAATA
- Phenotype
-
MIM: 612636
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
UNC80
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75640079] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- UNC80
- Gene Name
- unc-80 homolog, NALCN activator
View Full Product Details