Product Details

SNP ID
rs2302458
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1467248 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAGCAGGCCCCACTCACCACAGC[A/C]AGGTGCCCGTTCTTGGCCTTGGTGA
Phenotype
MIM: 606538
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MYO1C PubMed Links

Gene Details

Gene
MYO1C
Gene Name
myosin IC
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080779.1 3163 Silent Mutation CTG,CTT L1053L NP_001074248.1
NM_001080950.1 3163 Silent Mutation CTG,CTT L1034L NP_001074419.1
NM_033375.4 3163 Silent Mutation CTG,CTT L1018L NP_203693.3

View Full Product Details