Product Details

SNP ID
rs2302618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:29566359 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGACAGCGGCGATGCTGGCAGCTC[C/G]GGCCCCTGGTGGAAATCGCTCACCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRR15 PubMed Links

Gene Details

Gene
PRR15
Gene Name
proline rich 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175887.2 1545 Silent Mutation TCC,TCG S10S NP_787083.1
XM_005249653.3 1545 Silent Mutation TCC,TCG S10S XP_005249710.1
XM_011515198.1 1545 Silent Mutation TCC,TCG S10S XP_011513500.1
XM_011515199.1 1545 Silent Mutation TCC,TCG S10S XP_011513501.1
XM_017011842.1 1545 Silent Mutation TCC,TCG S10S XP_016867331.1

View Full Product Details