Product Details

SNP ID
rs2311230
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:26999653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCATTCTAACTGCTCAGGCTAG[G/T]GGATCCCAGCCAAGGACAAAGGCCC
Phenotype
MIM: 616824
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM46B PubMed Links

Gene Details

Gene
FAM46B
Gene Name
family with sequence similarity 46 member B
There are no transcripts associated with this gene.

Gene
TRNP1
Gene Name
TMF1-regulated nuclear protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013642.2 Intron NP_001013664.1
XM_005245867.3 Intron XP_005245924.1

View Full Product Details