Product Details

SNP ID
rs2101092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:173394762 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACACTATGTTATTTGATGTCACA[A/G]TTTACATCTATTTATATTGTGTATC
Phenotype
MIM: 603163
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCRG1 PubMed Links

Gene Details

Gene
SCRG1
Gene Name
stimulator of chondrogenesis 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011531578.1 Intron XP_011529880.1
XM_017007678.1 Intron XP_016863167.1

View Full Product Details