Product Details
- SNP ID
-
rs2797523
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
61
- Location
-
Chr.1:110661563 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCATTTTATTTCCTAAGTGCCTAGG[C/T]TCTGCTCAATGCTACTGCAGCTTTG
- Phenotype
-
MIM: 176263
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KCNA3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs151082169] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KCNA3
- Gene Name
- potassium voltage-gated channel subfamily A member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002232.4 |
|
Intron |
|
|
NP_002223.3 |
View Full Product Details