Product Details

SNP ID
rs2841225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104869373 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGCAGGGAGCTGCCCTGCCCACTC[A/G]GACGTTGCCTCCTGGATGCAACGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP170B PubMed Links

Gene Details

Gene
CEP170B
Gene Name
centrosomal protein 170B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112726.2 807 Intron NP_001106197.1
NM_015005.2 807 Intron NP_055820.2
XM_005267550.3 807 Intron XP_005267607.1
XM_011536665.2 807 Intron XP_011534967.1
XM_011536666.2 807 Intron XP_011534968.1
XM_011536667.2 807 Intron XP_011534969.1
XM_017021227.1 807 Silent Mutation TCA,TCG S20S XP_016876716.1
XM_017021228.1 807 Silent Mutation TCA,TCG S20S XP_016876717.1
XM_017021229.1 807 Silent Mutation TCA,TCG S20S XP_016876718.1
XM_017021230.1 807 Silent Mutation TCA,TCG S20S XP_016876719.1

View Full Product Details