Product Details

SNP ID
rs2838772
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:45081744 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCCTCTCTGAGTGCGATGTGGTA[C/T]GCTGAGGCCCAGAGGGAAAGCATGA
Phenotype
MIM: 601218
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADARB1 PubMed Links

Gene Details

Gene
ADARB1
Gene Name
adenosine deaminase, RNA specific B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112.3 Intron NP_001103.1
NM_001160230.1 Intron NP_001153702.1
NM_015833.3 Intron NP_056648.1
NM_015834.3 Intron NP_056649.1
XM_006723954.2 Intron XP_006724017.1
XM_011529425.2 Intron XP_011527727.1
XM_011529432.1 Intron XP_011527734.1
XM_017028242.1 Intron XP_016883731.1
XM_017028243.1 Intron XP_016883732.1
XM_017028244.1 Intron XP_016883733.1
XM_017028245.1 Intron XP_016883734.1
XM_017028246.1 Intron XP_016883735.1
XM_017028247.1 Intron XP_016883736.1
XM_017028248.1 Intron XP_016883737.1
XM_017028249.1 Intron XP_016883738.1
XM_017028250.1 Intron XP_016883739.1
XM_017028251.1 Intron XP_016883740.1
XM_017028252.1 Intron XP_016883741.1
XM_017028253.1 Intron XP_016883742.1
XM_017028254.1 Intron XP_016883743.1
XM_017028255.1 Intron XP_016883744.1
XM_017028256.1 Intron XP_016883745.1
Gene
LOC105372836
Gene Name
uncharacterized LOC105372836
There are no transcripts associated with this gene.

Gene
SSR4P1
Gene Name
signal sequence receptor subunit 4 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details