Product Details

SNP ID
rs2178395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54344066 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTTATCTGATGTGGTTCAACCTA[A/G]TAACTTTCTCAGGGTCACACATGTA
Phenotype
MIM: 615472 MIM: 613787
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COPZ1 PubMed Links

Gene Details

Gene
COPZ1
Gene Name
coatomer protein complex subunit zeta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271734.1 Intron NP_001258663.1
NM_001271735.1 Intron NP_001258664.1
NM_001271736.1 Intron NP_001258665.1
NM_016057.2 Intron NP_057141.1
XM_017018997.1 Intron XP_016874486.1
Gene
LOC102724050
Gene Name
uncharacterized LOC102724050
There are no transcripts associated with this gene.

Gene
MIR148B
Gene Name
microRNA 148b
There are no transcripts associated with this gene.

View Full Product Details