Product Details

SNP ID
rs2914967
Assay Type
Functionally tested
NCBI dbSNP Submissions
39
Location
Chr.1:25964254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAACAATACAGAGATGAGCACGGC[C/G]CTTGCACAAGGATGACATGCAAATT
Phenotype
MIM: 602344
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PAFAH2 PubMed Links

Gene Details

Gene
PAFAH2
Gene Name
platelet activating factor acetylhydrolase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000437.3 Intron NP_000428.2
XM_006710669.3 Intron XP_006710732.1
XM_006710670.3 Intron XP_006710733.1
XM_011541528.2 Intron XP_011539830.1
XM_011541529.2 Intron XP_011539831.1
XM_017001390.1 Intron XP_016856879.1
XM_017001391.1 Intron XP_016856880.1
XM_017001392.1 Intron XP_016856881.1

View Full Product Details