Product Details

SNP ID
rs2905755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29792596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCGCGGGCGCCGTGGATGGAGC[C/A]GGAGGGGCCGGAATATTGGGAAGAG
Phenotype
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
HCG4 PubMed Links

Gene Details

Gene
HCG4
Gene Name
HLA complex group 4 (non-protein coding)
There are no transcripts associated with this gene.

Gene
LOC554223
Gene Name
histocompatibility antigen-related
There are no transcripts associated with this gene.

View Full Product Details