Product Details

SNP ID
rs2071600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41477428 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAAGAGAACAGAACTCCAGACTCC[A/G]AATGCCAGGCTCTTTCTAGGCACCA
Phenotype
MIM: 602760 MIM: 602764 MIM: 604540
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT32 PubMed Links

Gene Details

Gene
KRT32
Gene Name
keratin 32
There are no transcripts associated with this gene.

Gene
KRT35
Gene Name
keratin 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002280.4 Intron NP_002271.3
XM_011524794.1 Intron XP_011523096.1
Gene
KRT36
Gene Name
keratin 36
There are no transcripts associated with this gene.

View Full Product Details