Product Details

SNP ID
rs2068247
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:53099480 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTCCTGTTGTGTCAGCCTTGTTCA[T/C]AGCAAGGGAACAATATATCTGAGCT
Phenotype
MIM: 192974 MIM: 603708
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
ITGA2 PubMed Links

Gene Details

Gene
ITGA2
Gene Name
integrin subunit alpha 2
There are no transcripts associated with this gene.

Gene
MOCS2
Gene Name
molybdenum cofactor synthesis 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004531.4 Intron NP_004522.1
NM_176806.3 Intron NP_789776.1

View Full Product Details