Product Details

SNP ID
hCV15862443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7217501 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAACTGGATTTCGGGGAGCCCC[C/G]GGGTAGGGGGGGGTCTTGCCAAACG
Phenotype
MIM: 609575 MIM: 602887 MIM: 602151
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACADVL PubMed Links

Gene Details

Gene
ACADVL
Gene Name
acyl-CoA dehydrogenase, very long chain
There are no transcripts associated with this gene.

Gene
DLG4
Gene Name
discs large MAGUK scaffold protein 4
There are no transcripts associated with this gene.

Gene
DVL2
Gene Name
dishevelled segment polarity protein 2
There are no transcripts associated with this gene.

Gene
MIR324
Gene Name
microRNA 324
There are no transcripts associated with this gene.

View Full Product Details