Product Details
- SNP ID
-
rs2240593
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:15744409 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACCTTCCTGCCCAAAAAAGGAGGAA[C/T]GAGGGTCAACCAGCAGCAGACCAGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TBC1D26
PubMed Links
Gene Details
- Gene
- TBC1D26
- Gene Name
- TBC1 domain family member 26
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178571.4 |
1494 |
UTR 3 |
|
|
NP_848666.2 |
View Full Product Details