Product Details

SNP ID
rs2240728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:22812880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAGGCTTAAATAGCACCTAGGTA[C/T]ACACATAAATTCCCTGTTTTGAGAA
Phenotype
MIM: 607980
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TOMM7 PubMed Links

Gene Details

Gene
TOMM7
Gene Name
translocase of outer mitochondrial membrane 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019059.3 531 UTR 3 NP_061932.1

View Full Product Details