Product Details

SNP ID
rs2241091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3198284 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTTATATTATGGTACAACGATG[G/T]GCATGTATTTCCGCCCTCTGACCAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR1A2 PubMed Links

Gene Details

Gene
OR1A2
Gene Name
olfactory receptor family 1 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012352.2 768 Missense Mutation GGC,TGC G256C NP_036484.1

View Full Product Details