Product Details

SNP ID
rs2241093
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:3197533 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTGAAGAAGCCATGAAGAAAGAAAA[C/T]CAATCCTTTAACCTGGATTTTATTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR1A2 PubMed Links

Gene Details

Gene
OR1A2
Gene Name
olfactory receptor family 1 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012352.2 17 Silent Mutation AAC,AAT N5N NP_036484.1

View Full Product Details