Product Details
- SNP ID
-
rs2269330
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:4385253 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GGTTTGTGGCTGAGAAGCGGTGCCA[A/G]TGGGGAGAGTGGCAGTGTCCAGAGA
- Phenotype
-
MIM: 109092
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR52B3P
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74051867] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR52B3P
- Gene Name
- olfactory receptor family 52 subfamily B member 3 pseudogene
There are no transcripts associated with this gene.
- Gene
- TRIM21
- Gene Name
- tripartite motif containing 21
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003141.3 |
1573 |
UTR 3 |
|
|
NP_003132.2 |
View Full Product Details