Product Details

SNP ID
rs2230196
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:156309177 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GGGGAGACTCTGCTGGTTCTGTGCA[G/T]TGAAGTAGCCTTGCCTAGCACTCAC
Phenotype
MIM: 600114
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCT3 PubMed Links

Gene Details

Gene
CCT3
Gene Name
chaperonin containing TCP1 subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008800.2 1777 UTR 3 NP_001008800.1
NM_005998.4 1777 UTR 3 NP_005989.3
Gene
VHLL
Gene Name
von Hippel-Lindau tumor suppressor like
There are no transcripts associated with this gene.

View Full Product Details