Product Details

SNP ID
rs2275764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:118960534 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCCTCTCTTTCAGCACCTTTTCT[C/G]TTTTAGCTGATCGTACAGATGTGTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM184A PubMed Links

Gene Details

Gene
FAM184A
Gene Name
family with sequence similarity 184 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100411.2 Intron NP_001093881.1
NM_001288576.1 Intron NP_001275505.1
NM_024581.5 Intron NP_078857.5

View Full Product Details