Product Details

SNP ID
rs2288887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51992550 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTGTGAACTCTCTCAATCTGTTGA[A/G]GTTTTATTTTTGGGCAAAGACGTTC
Phenotype
MIM: 605422
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF350 PubMed Links

Gene Details

Gene
ZNF350
Gene Name
zinc finger protein 350
There are no transcripts associated with this gene.

Gene
ZNF615
Gene Name
zinc finger protein 615
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199324.1 2876 UTR 3 NP_001186253.1
NM_001321317.1 2876 UTR 3 NP_001308246.1
NM_001321318.1 2876 UTR 3 NP_001308247.1
NM_001321319.1 2876 UTR 3 NP_001308248.1
NM_001321320.1 2876 UTR 3 NP_001308249.1
NM_001321321.1 2876 UTR 3 NP_001308250.1
NM_001321322.1 2876 UTR 3 NP_001308251.1
NM_001321323.1 2876 UTR 3 NP_001308252.1
NM_198480.3 2876 UTR 3 NP_940882.3
XM_011526824.1 2876 UTR 3 XP_011525126.1
XM_011526825.1 2876 UTR 3 XP_011525127.1
XM_011526826.1 2876 UTR 3 XP_011525128.1
XM_011526829.1 2876 UTR 3 XP_011525131.1
XM_011526830.1 2876 UTR 3 XP_011525132.1
XM_017026648.1 2876 UTR 3 XP_016882137.1
XM_017026649.1 2876 UTR 3 XP_016882138.1
XM_017026650.1 2876 UTR 3 XP_016882139.1
XM_017026651.1 2876 UTR 3 XP_016882140.1

View Full Product Details