Product Details

SNP ID
rs2276004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:37937015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCACTTGGTGTCAACTTGCAGT[C/G]GATAATGTTCCCCATCACAAGGACA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GOT1L1 PubMed Links

Gene Details

Gene
GOT1L1
Gene Name
glutamic-oxaloacetic transaminase 1-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152413.2 1031 Missense Mutation CAC,GAC H188D NP_689626.2
XM_005273399.3 1031 Missense Mutation CAC,GAC H187D XP_005273456.1
XM_006716285.3 1031 Intron XP_006716348.1
XM_011544391.2 1031 Missense Mutation CAC,GAC H6D XP_011542693.1

View Full Product Details