Product Details

SNP ID
rs2289194
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:25236758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACCAGGGAGGAAGGGCTGAAGG[C/T]CAGCAGCCCTGGGCCCTCCTCTGGC
Phenotype
MIM: 602769
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DNMT3A PubMed Links

Gene Details

Gene
DNMT3A
Gene Name
DNA methyltransferase 3 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320892.1 Intron NP_001307821.1
NM_001320893.1 Intron NP_001307822.1
NM_022552.4 Intron NP_072046.2
NM_153759.3 Intron NP_715640.2
NM_175629.2 Intron NP_783328.1
NM_175630.1 Intron NP_783329.1
XM_005264175.4 Intron XP_005264232.1
XM_005264177.4 Intron XP_005264234.1
XM_011532662.2 Intron XP_011530964.1
XM_011532663.2 Intron XP_011530965.1
XM_011532664.2 Intron XP_011530966.1
XM_011532666.2 Intron XP_011530968.1
XM_011532667.2 Intron XP_011530969.1
XM_017003526.1 Intron XP_016859015.1
XM_017003527.1 Intron XP_016859016.1

View Full Product Details