Product Details

SNP ID
hCV15881150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44518690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCCTGCCCCCCACCAGCTCATT[C/T]GTTAGATGATTGGAAGTGACTGCCT
Phenotype
MIM: 612920
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TSPEAR PubMed Links

Gene Details

Gene
TSPEAR
Gene Name
thrombospondin type laminin G domain and EAR repeats
There are no transcripts associated with this gene.

Gene
TSPEAR-AS1
Gene Name
TSPEAR antisense RNA 1
There are no transcripts associated with this gene.

Gene
TSPEAR-AS2
Gene Name
TSPEAR antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details