Product Details

SNP ID
rs2980086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:4273140 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTTGGGGAGGAGCGAAAGGGAAC[A/C]GGGTCTCAGAAAGCTATATTGGGCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LYAR PubMed Links

Gene Details

Gene
LYAR
Gene Name
Ly1 antibody reactive
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145725.1 Intron NP_001139197.1
NM_017816.2 Intron NP_060286.1
XM_011513505.1 Intron XP_011511807.1
XM_011513506.2 Intron XP_011511808.1

View Full Product Details