Product Details

SNP ID
rs2289807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:48430976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGAAACAGTGAATTACAGGAAA[C/G]ATTAGTACTTCCTTTTAATATGATC
Phenotype
MIM: 610585 MIM: 601158
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP22 PubMed Links

Gene Details

Gene
ARHGAP22
Gene Name
Rho GTPase activating protein 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256024.1 Intron NP_001242953.1
NM_001256025.2 Intron NP_001242954.1
NM_001256026.1 Intron NP_001242955.1
NM_021226.3 Intron NP_067049.2
XM_005270014.3 Intron XP_005270071.1
XM_011540002.1 Intron XP_011538304.1
XM_011540003.1 Intron XP_011538305.1
XM_011540004.1 Intron XP_011538306.1
XM_011540005.1 Intron XP_011538307.1
XM_011540006.1 Intron XP_011538308.1
XM_011540007.2 Intron XP_011538309.1
XM_011540011.2 Intron XP_011538313.1
XM_011540012.2 Intron XP_011538314.1
XM_011540013.2 Intron XP_011538315.1
XM_011540015.2 Intron XP_011538317.1
XM_017016470.1 Intron XP_016871959.1
XM_017016471.1 Intron XP_016871960.1
XM_017016472.1 Intron XP_016871961.1
XM_017016473.1 Intron XP_016871962.1
XM_017016474.1 Intron XP_016871963.1
XM_017016475.1 Intron XP_016871964.1
XM_017016476.1 Intron XP_016871965.1
XM_017016477.1 Intron XP_016871966.1
Gene
MAPK8
Gene Name
mitogen-activated protein kinase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278547.1 Intron NP_001265476.1
NM_001278548.1 Intron NP_001265477.1
NM_001323302.1 Intron NP_001310231.1
NM_001323320.1 Intron NP_001310249.1
NM_001323321.1 Intron NP_001310250.1
NM_001323322.1 Intron NP_001310251.1
NM_001323323.1 Intron NP_001310252.1
NM_001323324.1 Intron NP_001310253.1
NM_001323325.1 Intron NP_001310254.1
NM_001323326.1 Intron NP_001310255.1
NM_001323327.1 Intron NP_001310256.1
NM_001323328.1 Intron NP_001310257.1
NM_001323329.1 Intron NP_001310258.1
NM_001323330.1 Intron NP_001310259.1
NM_001323331.1 Intron NP_001310260.1
NM_139046.3 Intron NP_620634.1
NM_139049.3 Intron NP_620637.1

View Full Product Details